VWR® Rubber Bulbs
Catalog # 15001-364
Supplier: VWR International
Specifications
- Capacity:2 ml
- Color:Red
- Cat. no.:15001-364
Specifications
About this item
Made of durable natural rubber, bulbs are suitable for medicine droppers and small dropping pipettes.
- Durable
- Natural rubber construction
- Heavy-walled design
- Cylindrical
Neck is grooved to take flat flange of pipettes. Bulbs fit pipettes 6 to 8 mm ext. Ø.
Customers Who Bought This Also Bought