VWR® Disposable Pasteur Pipettes, Glass, Plugged
# de catalogue 14672-400
Fournisseur: VWR International
Spécifications
- Material:Borosilicate glass, 7.0 expansion
- Cat. no.:14672-400
- Overall Length:14.6 cm (5¾")
Spécifications
A propos de cet article
These disposable glass pipettes are furnished with cotton plugs.
- Disposable
- Glass fabrication
- Plugged
A positive constriction at the top holds plug in place.
Customers Who Bought This Also Bought