ContactUsLinkComponent
VWR* Tissue Wipers Dispenser
VWR® Dispensers for VWR® Light-Duty Tissue Wipers
# de catalogue 14233-760
Fournisseur:  VWR International
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
VWR* Tissue Wipers Dispenser
VWR® Dispensers for VWR® Light-Duty Tissue Wipers
# de catalogue 14233-760
Fournisseur:  VWR International
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
  • Description:
    Dispenser, Large, for 35.6W x 42.4L cm (14 x 163/4") Wipers
Spécifications
A propos de cet article
White PVC dispensers for tissue boxes. Can be wall-mounted or placed on a bench.
  • Hold wiper box securely, providing one-handed access
  • Double-faced tape allows semi-permanent installation
  • Features keyhole openings for screws
Customers Who Bought This Also Bought