Finntip™ Pipette Tips, Thermo Scientific
# de catalogue 14232-916
Fournisseur: Thermo Fisher Scientific
Spécifications
- Volume:100 - 1000 µL
- Packed:10×96/Rack
- Sterile:+
- Tip Style:Finntip™ 1000
- Supplier No.:9401113
Spécifications
A propos de cet article
These high-quality pipette tips are specifically designed to fit Finnpipette® pipettors, but are also compatible with most pipettor brands.
- Made of high-quality polypropylene
- Autoclavable
- Chemical-resistant
Tips in refill trays can be used to quickly refill existing racks.
Customers Who Bought This Also Bought