PCR Plate Sealers, Sorenson BioScience
# de catalogue 14230-062
Fournisseur: Sorenson Bioscience
Spécifications
- Sterility:Non sterile
- Description:Sealer Plate Aluminum
- Supplier No.:36890
Spécifications
A propos de cet article
Adhesive seals and cap strips prevent evaporation during thermal cycling or storage and are compatible with Sorenson BioScience PCR plates and tube strips.
Certified free of RNase / DNase, Human DNA, Pyrogens and PCR Inhibitors.
Customers Who Bought This Also Bought