Accessories for Domed Lid Visible Range Spectrophotometer, Model 6320D, Techne
# de catalogue 14229-096
Fournisseur: Argos Technologies
Spécifications
- Description:Visible Glass Cuvette, 334–2500 nm, 20 x 10 mm, 7 mL
- Cat. no.:14229-096
- Supplier No.:035-086
Spécifications
A propos de cet article
Customers Who Bought This Also Bought