Finntip Stepper Tips, Thermo Scientific
# de catalogue 14222-046
Fournisseur: Thermo Fisher Scientific
Spécifications
- Volume:100 µl, 200 µl, 300 µl, 400 µl, 500 µl
- Sterile:+
- Tip Style:Finntip Stepper 5.0 mL
- Supplier No.:9404203
Spécifications
A propos de cet article
Finntip stepper tips are the perfect fit with Thermo Scientific Finnpipette stepper pipetters.
The finntip stepper tips are plunger-style tips for positive displacement pipetting. Seven sizes are available in sterile and non sterile versions.
Customers Who Bought This Also Bought