VWR® High Strength Autoclavable Infectious Waste Bag
# de catalogue 14220-100
Fournisseur: VWR International
Small Business Enterprise
Spécifications
- W×L:305×610 mm (12×24")
- Color:Red, plain
- Thickness:2 mil
Spécifications
A propos de cet article
These 2 mil polypropylene bags are engineered to meet the most demanding regulatory specifications worldwide.
- Designed to handle dangerous materials
- High-strength
- Autoclavable
They exceed the 480 gram tear test as defined by ASTM D1922 in both length (MD) and width (TD) directions.
These high strength bags are ideal for hazardous waste collection and decontamination in an autoclave. Bags are formulated to withstand stress at the side seams and are strong enough to hold items in the laboratory.
These bags are red and are available printed with a black biohazard symbol or plain. The printed bags also read 'Infectious Waste' and feature a temperature indicator patch. All bags are autoclavable to 135 °C (275 °F).
Certifications: Bags meet all state requirements for hazardous waste handling, including Florida, Pennsylvania and California Department of Health codes. ASTM D1922 for tear resistance greater than 480 grams in both width and height of bag.
Ordering information: Bags are supplied with 12.7 cm (5) twist ties.
Customers Who Bought This Also Bought