ContactUsLinkComponent
Condenser, Allihn, for Soxhlet Extraction Apparatus
SP Wilmad-LabGlass Allihn Condensers, Soxhlet, SP Industries
Catalog # 14209-840
Supplier:  SP Industries
Condenser, Allihn, for Soxhlet Extraction Apparatus
SP Wilmad-LabGlass Allihn Condensers, Soxhlet, SP Industries
Catalog # 14209-840
Supplier:  SP Industries
Specifications
  • ST Joint:
    34/45
  • Description:
    Allihn Condenser
  • Cat. no.:
    14209-840
  • Bottom [ST] Joint Size:
  • Inner Diameter:
  • Supplier No.:
    LG-6900-110
Specifications
About this item
Allihn condensers are designed for use within a Soxhlet extraction apparatus.
Customers Who Bought This Also Bought