SP Wilmad-LabGlass Carboys with Hose Connection, SP Industries
# de catalogue 14207-556
Fournisseur: SP Industries
Spécifications
- Capacity:9.5 L (2.5 gal.)
- Cat. no.:14207-556
- Connectors:⅜" Hose Connection
- Supplier No.:LG-3465-100
Spécifications
A propos de cet article
Large capacity borosilicate glass bottles, offered in four different sizes, feature a hose connector sealed at neck (¹/₂ or ³/₈" connection).
All bottle necks are compatible with a #12 stopper.
Customers Who Bought This Also Bought