ContactUsLinkComponent
Stalagmometer, Tube Only
SP Wilmad-LabGlass Tube for Stalagmometer, SP Industries
# de catalogue 14203-776
Fournisseur:  SP Industries
Stalagmometer, Tube Only
SP Wilmad-LabGlass Tube for Stalagmometer, SP Industries
# de catalogue 14203-776
Fournisseur:  SP Industries
Spécifications
  • Capacity:
    2.5 ml
  • Description:
    Stalagmometer Tube
  • Cat. no.:
    14203-776
  • Supplier No.:
    LG-5050-102
Spécifications
A propos de cet article
Specifically designed for checking the surface tension of liquids, each tube is accurately calibrated to quickly determine surface tension by the drop count method.
  • Graduated above and below the bulb
This tube allow for flexibility in sample size of most liquid baths.
Delivery: All tubes are supplied with instructions for use.
Customers Who Bought This Also Bought