ContactUsLinkComponent
Flask, Filter, Heavy Wall, [ST] Joint
SP Wilmad-LabGlass Filterware™ Filter Flask, SP Industries
Catalog # 14203-552
Supplier:  SP Industries
SP Wilmad-LabGlass Filterware™ Filter Flask, SP Industries
Catalog # 14203-552
Supplier:  SP Industries
Specifications
  • ST Joint No.:
    40/35
  • Capacity:
    2000 ml
  • Description:
    Filter Flask
  • Cat. no.:
    14203-552
  • Supplier No.:
    BP-1827-002
Specifications
About this item
These microfiltration components are designed for the filtration of aqueous liquids
  • HPLC mobile phase filtration
  • Suitable for general laboratory microfiltration
The components are interchangeable with competitive product offerings and can be used with filtering manifolds.
They may be used for analysis of particulate or bacterial contamination.
Customers Who Bought This Also Bought