SP Wilmad-LabGlass Chromatography TLC Sprayers, SP Industries
Catalog # 14008-128
Supplier: SP Industries
Specifications
- Capacity:125 ml
- Description:TLC Sprayer, Complete Assembly
- Cat. no.:14008-128
- ST Joint No.:24/40
- Supplier No.:LG-4590-100
Specifications
About this item
When using thin layer or paper chromatography (TLC), these borosilicate glass atomizers are used for spraying chromogenic reagents.
- The air inlet tube features an opening to release pressure and stop spraying
- All joints are Standard Taper 24/40
Ordering information: Complete unit consists of the spray head, flask and LG-1004 clamp.
Customers Who Bought This Also Bought