Automation Pipette Tips for Tecan Te-MO and Freedom EVO Workstations, Thermo Scientific
14004-624
- Volume:50 µl
- Tip Color:Clear
- Packed:96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case
- Cat. no.:14004-624
- Tip Style:Standard
- For Use With:Tecan automated workstations
- Supplier No.:163-96R
Thermo Scientific provides a solution for your Tecan robotic pipette tips.
These automation pipette tips have been designed, validated, and tested on a Tecan EVO robot to ensure equal performance without the need to modify your existing protocols and programs. Our low percentage Coefficient of Variation (%CV) ensures product consistency and performance from every lot. Each lot must pass a 15-point QC inspection including performance testing on our Tecan workstation before tips are released. All tips are certified RNase, DNase, DNA, endotoxin, bioburden, and pyrogen free.
Frequently Bought Together