Specifications
- Description:ROSS pH electrode storage solution
- Size:475 ml bottle
- Cat. no.:14004-272
- Supplier No.:810001
Specifications
About this item
Cleaning Solutions restore proper electrode performance and prolong the useful life of the electrode.
- Restore electrode performance
- Prolong usefulness
- Unique formulations
Cleaning Solutions are identified by the letters A, B, C, and D. Each letter designates a unique formulation. Cleaning solution A is for removing protein contaminants. Cleaning solution B is for removing bacterial contaminants. Cleaning solution C is for general cleaning. Cleaning solution D is for removing oil and grease contaminants. Bottles include a small beaker to hold the solution and a plastic pipet to remove the old electrode filling solution.