Pipette Tips, Sorenson BioScience
# de catalogue 13503-464
Fournisseur: Sorenson Bioscience
Spécifications
- Volume:100 - 1000 µl
- Tip Color:Blue
- Cat. no.:13503-464
- Tip Style:MicroReach
- Supplier No.:27120
Spécifications
A propos de cet article
Sorenson BioScience universal fit pipette tips are manufactured from medical grade polypropylene in a tightly controlled production environment.
Certified free of RNase / DNase, Human DNA, Pyrogens and PCR Inhibitors.
The accurate and precise tips are designed to fit most research-grade pipettors. Available in low binding, pipette tips reduce sample retention.
Customers Who Bought This Also Bought