DuPont™ Coveralls with Elastic Wrists/Ankles, Magid Glove
# de catalogue 13450-180
Fournisseur: Magid Glove
Small Business Enterprise
Spécifications
- Clothing Size:3XL
- Garment application:Industrial
- Sterile:+
- Color:White
- Description:Coveralls with Elastic Wrists/Ankles
- Fabric:Tyvek
- Cat. no.:13450-180
- Supplier No.:CVC11XXXL
Spécifications
A propos de cet article
Zippered coveralls are made of Tyvek, a tough polyolefin material which provides the best balance of protection, durability and comfort.
- Elastic border at wrists and ankles
- Disposable
Greater range of movement while stretching and bending. Tailored fit and reinforcement in high stress areas. Tear and puncture resistant garments that withstand most chemicals and acids.
Customers Who Bought This Also Bought