DuPont™ Tyvek® Coveralls with Attached Hood, Magid Glove
# de catalogue 13450-166
Fournisseur: Magid Glove
Small Business Enterprise
Spécifications
- Clothing Size:L
- Garment application:Industrial,Laboratory
- Color:White
- Description:DuPont™ Tyvek® Coveralls with Attached Hood
- Seam type:Serged
- Cat. no.:13450-166
- Supplier No.:CVCH11L
Spécifications
A propos de cet article
Disposable coveralls are made of Tyvek®, a tough polyolefin material which provides the best balance of protection, durability, and comfort.
- Zippered front
- Remains strong, wet or dry
- Elastic border at wrists and ankles
- Comfortable and more tailored fit
These lightweight overalls feature a great range of movement while stretching and bending, improved mobility, a tailored fit, and reinforcement in high stress areas.
Coveralls are ideal for use against such non-hazardous materials as dirt and grime, animal waste, paints, oil and grease, lubricants, or hazardous materials including fertilizer, pesticides, asbestos, lead, chromium, mold, fiberglass, and radioactive particles.
Customers Who Bought This Also Bought