VWR® Modular Heating Blocks for Standard Test Tubes
# de catalogue 13259-162
Fournisseur: VWR International
Product available on GSA Advantage®
Spécifications
- Description:Block
- No. of wells:12
- Cat. no.:13259-162
- For Use With:Standard Test Tubes
- For Tube Diameter:15/16 mm
- Well Diameter:17.5 mm
Spécifications
A propos de cet article
These blocks have been designed for standard test tubes.
- Now with Best-in-Class, Exclusive 5-Year Warranty
Constructed from a solid anodized aluminum block, the close contact of tubes to block walls allows for maximum heat retention. Single block size.
Customers Who Bought This Also Bought