VWR® Talon® Support Stands, Cast Iron, Rectangular Base
# de catalogue 12985-070
Fournisseur: VWR International
Product available on GSA Advantage®
Spécifications
- Dimensions:12.7 x 20.3 cm (5 x 8")
- Cat. no.:12985-070
Spécifications
A propos de cet article
Support stand bases are constructed of cast iron with black enamel finish.
- Cast iron construction with black enamel finish
- Built-in support rod holder
- Rectangular base
- For use with VWR® Talon® lab frames
All bases feature a built-in support rod holder with locking knob that accepts a 13mm (1/2") diameter support rod (sold separately).
Customers Who Bought This Also Bought