Spécifications
- Cat. no.:12620-900
- Maximum Load:30 kg (66 lbs.)
- Vertical Lift:6.4–12.7 cm (21/2–5")
- Platform Size:10.2×10.2 cm (4×4")
Spécifications
A propos de cet article
Aluminum lab jacks provide stable height adjustment for various items in the labs such as flasks, baths, and small equipment.
- Exceptional stability and durability
- Aluminum construction
- Four convenient sizes
Oversized, sure-grip adjustment knobs provide smooth and accurate height adjustment
Support rod kit includes a 43.2 cm (17") vertical support rod for mounting to the upper deck of a 15.2x15.2 cm (6x6") or 20.3x20.3 cm (8x8") support jack. Ideal for creating an adjustable support stand for mounting various items such as thermometer clamps, temperature probes, or flask and column clamps. Kit also includes a jam nut and a flat washer.
Top and bottom decks are constructed of anodized aluminum. Internal supports and drive screw are constructed of stainless steel. Lab jacks are designed to hold the specified weight after it is lifted to the preferred height.