Titration Cup, Dyn-A-Med
Catalog # 12578-108
Supplier: Dyn-A-Med
Specifications
- Capacity:100 ml (3.4 oz.)
- Description:Titration Cup
- Cat. no.:12578-108
- Supplier No.:80099
Specifications
About this item
Made of polypropylene.
Graduated on two sides and has a writing surface. For METTLER TOLEDO® DL series titrators.
Customers Who Bought This Also Bought