VWR® Aluminum Micro Weighing Dishes
Catalogue # 12577-068
Fournisseur: VWR International
Spécifications
- Capacity:1.12 ml
- Description:Oval Weighing Dishes
- Dimensions:25W x 7L x 7D mm
- Cat. no.:12577-068
Spécifications
À propos de cet article
Ideal for microbalances, organic micro elementary and thermal analysis, and pharmaceutical use.
- Easy to use
- Compatible with most commercially available analyzers and microbalances
- For use in a variety of applications
Our micro weighing boats are compatible with most commercially available analyzers and microbalances. Packaged in easy-to-use plastic containers for storage and dispensing.
Frequently Bought Together