ContactUsLinkComponent
Novus Pipettor Stand, Thermo Scientific
Novus Pipettor Stand, Thermo Scientific
# de catalogue 11388-556
Novus Pipettor Stand, Thermo Scientific
Novus Pipettor Stand, Thermo Scientific
# de catalogue 11388-556
Spécifications
  • Description:
    Novus Pipettor Stand
  • Cat. no.:
    11388-556
  • Supplier No.:
    9420360
Spécifications
A propos de cet article
This easy-to-use mechanical stand enables pipettors to be stored safely in an upright position
The stand comfortably accommodates all single and multichannel Finnpipette* Novus and Focus pipettors. Its compact size ensures maximum convenience and safety in the laboratory.
Customers Who Bought This Also Bought