ContactUsLinkComponent
FINNTIP FLEX 300UL EA960 CS960
Finntip™ Pipette Tips, Thermo Scientific
# de catalogue 11388-532
Finntip™ Pipette Tips, Thermo Scientific
# de catalogue 11388-532
Spécifications
  • Volume:
    5 - 300 µl
  • Packed:
    10×96/Rack
  • Tip Style:
    Finntip™ Flex 300
  • Supplier No.:
    94060510
Spécifications
A propos de cet article
These high-quality pipette tips are specifically designed to fit Finnpipette® pipettors, but are also compatible with most pipettor brands.
  • Made of high-quality polypropylene
  • Autoclavable
  • Chemical-resistant
Tips in refill trays can be used to quickly refill existing racks.
Customers Who Bought This Also Bought