Septa, Fluoropolymer Resin Silicone, Qorpak®
# de catalogue 11311-628
Fournisseur: Qorpak
Small Business Enterprise
Spécifications
- Ø:20 mm
- Color:White
- Thickness:0.125”
- Cat. no.:11311-628
- Supplier No.:CAP-00563
Spécifications
A propos de cet article
Resist coring and provide great resealability.
Ideal for when multiple injections are required. Septa maintain PTFE resistance until punctured, then have silicone compatibility.
Customers Who Bought This Also Bought