Infecon® Absorbent Strips, Com-Pac
Catalog # 11217-524
Supplier: Com-Pac
Specifications
- L×W:7.6×7.6 cm (3×3")
- Cat. no.:11217-524
- Absorbency:50 ml
- Supplier No.:INF-2003
Specifications
About this item
Designed for use with Infecon® shippers and diagnostic packaging, these absorbent strips provide fast and reliable protection against spills.
Frequently Bought Together