ContactUsLinkComponent
Vessel Hold-Down Clamp
Accessories for Dissolution Vessels and Centering System Accessories, Quality Lab Accessories
# de catalogue 11010-510
Fournisseur:  QLA
Vessel Hold-Down Clamp
Accessories for Dissolution Vessels and Centering System Accessories, Quality Lab Accessories
# de catalogue 11010-510
Fournisseur:  QLA
Spécifications
  • Description:
    Vessel Retainer Assembly for Distek 2100 and 6300 baths
  • Cat. no.:
    11010-510
  • Supplier No.:
    VSCLIP-DK