ContactUsLinkComponent
IsoTip Pipet Tips
Corning® IsoTip™ Pipette Tips, Corning
# de catalogue 10010-364
Fournisseur:  Corning
   Low Manufacturing Impact
Product available on GSA Advantage®
Corning® IsoTip™ Pipette Tips, Corning
# de catalogue 10010-364
Fournisseur:  Corning
   Low Manufacturing Impact
Product available on GSA Advantage®
Spécifications
  • Volume:
    0.1 - 2 µL
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    I-RECs, ISO 14001 - Reynosa
  • Packed:
    96/Rack
  • Cat. no.:
    10010-364
  • For Use With:
    Gilson® and Other Popular Ultra Micro Pipettors
  • Supplier No.:
    4801
Spécifications
A propos de cet article
These pipette tips feature an inert, hydrophobic bonded-fiber barrier that prevents aerosolised contaminants from coming in contact with air displacement pipettors. Ideal for applications where avoidance of cross contamination is critical, such as DNA amplification and radioisotope handling.
  • Exceptional durability
  • All Corning® tip racks stand up to the demands of use with multichannel pipettors and withstand repeated autoclaving cycles
  • Sterile and certified RNase-/ DNase-free and non pyrogenic
These pipette tips feature an inert, hydrophobic bonded-fiber barrier that prevents aerosolised contaminants from coming in contact with air displacement pipettors. Ideal for applications where avoidance of cross contamination is critical, such as DNA amplification and radioisotope handling.
Customers Who Bought This Also Bought