ESD Safe Spray Bottle, R & R Lotion
# de catalogue 56614-156
Fournisseur: R & R Lotion
Small Business Enterprise
Spécifications
- Capacity:473 ml (16 oz.)
- Description:ESD Safe Spray Bottle
- Cat. no.:56614-156
- Supplier No.:SCB-16-ESD
Spécifications
A propos de cet article
Designed for use around static sensitive electronics.
- Spray bottle with trigger
- Can be used with various cleaners
- Will not hold a charge
R&R Lotion I.C. antistatic dissipative ESD safe spray bottle comes in 16 ounce and 32 ounce sizes. Cleanroom blue static dissipative containers have an average surface resistivity of 10⁹ to 10¹⁰. All blue solvent dispensers will dissipate a charge of ±5000 volts in less than 2 s at 40 percent relative humidity. By their chemical nature, they will not have any chemical reactivity with solvents such as TCE or alcohol.
Certifications: Bottles help to meet ANSI/ESDA S20.20.
Customers Who Bought This Also Bought