ESD Safe Sports Bottle, R & R Lotion
# de catalogue 56616-120
Fournisseur: R & R Lotion
Small Business Enterprise
Spécifications
- Capacity:946 ml (32 oz.)
- Description:ESD Safe Sports Bottle
- Cat. no.:56616-120
- Supplier No.:SB-32-ESD
Spécifications
A propos de cet article
Antistatic dissipative, ESD safe sports bottle.I.C.
- Water/sports bottle in an ESD safe container
- Designed for use around static sensitive electronics
- Will not hold an electric charge
The ESD Safe Sports Bottle allows workers to keep drinks handy in static sensitive environments. Bottles feature a pull-top cap for workers on the go. These containers offer an average surface resistivity of 10⁹ to 10¹⁰. All blue solvent dispensers will dissipate a charge of +/- 5000 volts in less than 2 s at 40 percent relative humidity.
Customers Who Bought This Also Bought