A propos de cet article
Blue I.C. pre-gloving, anti-static moisturizing lotion helps relieve dermatitis symptoms.
- Greaseless and static dissipative
- Contains no Silicones, Lanolins, Mineral Oil, or Glycerins
- Non-contaminating
- Can be used before gloving
I.C
Because dry skin can reduce the conductivity of grounding wrist straps, and this lotion moisturizes skin without introducing electrically resistive ingredients, it can help to meet American National Standards Institute/Electrostatic Discharge Association (ANSI/ESDA) S20.20 requirements. The lotion is available in a variety of packaging options. It also contains botanical extracts of blueberry, cranberry, pomegranate, strawberry, raspberry, green coffee bean, green tea, grape seed, and acai berry.
Blue Lotion is static dissipative for use in cleanrooms and electrostatic protected areas. It is compatible with latex and nitrile gloves to reduce the risk of glove degradation, and with CHG (chlorhexidine gluconate), an anti-pathogen commonly used in hospitals to reduce the risk of deactivating interference. It does not contain glycerin, mineral oil, silicones, or lanolin, any of which may contaminate integrated circuit (I.C.) boards during manufacturing or handling. This lotion moisturizes dry skin with a combination of ingredients, including the humectant propylene glycol and vitamins A, D, and E, along with succulents such as aloe vera, jojoba, and blue agave. It is made with deionized water, absorbs immediately into the skin for a non-greasy feeling, and is fragrance-free to reduce the chance of allergic reactions.