3B Scientific® Spinal Cord And Nerve Endings Model
# de catalogue 470137-710
Fournisseur: American 3B Scientific
Spécifications
- Description:Spinal Cord And Nerve Endings
- Height:13 cm
- No. of Keyed Items:34
- Cat. no.:470137-710
- Length:26 cm
- Width:20 cm
- Supplier No.:C41
- No. of Parts:1
Spécifications
A propos de cet article
Great For Teaching Anatomy
- Buy Individually, Or As A Set
- Each Model Shows Basic Dental Structure3 Year Manufacturers Warranty
This spinal cord model illustrates the composition of the spinal cord, magnified to a scale of 5:1. The base of the model features illustrations of the various cross-sections of the spinal cord through the white and gray matter at the neck, torso, lumbar, and sacral regions. This model is supplied with a base.
Customers Who Bought This Also Bought