Franck-Hertz Experiment Power Supply
# de catalogue 470137-702
Fournisseur: American 3B Scientific
Spécifications
- Description:Franck-Hertz Experiment Power Supply
- Cat. no.:470137-702
- Supplier No.:U8482530-115
Spécifications
A propos de cet article
Adjustable Voltage Power Supply
This is the power supply unit for operating the mercury-filled Franck-Hertz tube and the neon-filled Franck-Hertz tube. It provides all the voltages needed to power the tubes and includes a sensitive built-in DC amplifier for measuring collector current. The accelerating voltage can be set-up manually on the apparatus or set to a saw-tooth wave form. Additional measuring inputs are also available for the anode current and accelerating voltage.
Customers Who Bought This Also Bought