FETFE O-Ring Replacement Sets for Stopcocks, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
FETFE replacement O-Ring sets are intended for use with stopcock plugs (see 89062-484 and 89062-576 series).
Ordering information: O-Ring sets 89062-502 and -504 include 18 sets per box. O-Ring set 89062-506 includes 12 sets per box. O-Ring set 89062-508 includes 6 sets per box.
Frequently Bought Together