Air Sampling Blower, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This shaded pole blower is designed to pull air through an air sampling manifold.
Blower has air delivery of 1.7 m3/min. (60 CFM) at 0.0 meters of water static pressure, which assures a minimum pressure drop across each port in the manifold train. Free air RPM is 3030. Operates on 115 volts, 50/60 cycles.
Ordering information: A 230 volt, 50 cycle version of this unit is available via special order. Contact your Avantor representative for details. For aluminum blower mount (not included), see 89059-254 (25 mm) or 89059-256 (50 mm) series.
Customers Who Bought This Also Bought