Float, Polyethylene, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This float is used on top of chromatography column packing to prevent turbulence caused by a drop-wise addition.
Customers Who Bought This Also Bought