Microscale Drying Tube, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Microscale drying tube is designed to protect moisture sensitive reaction components from atmospheric water vapor while allowing a reaction system to be kept unsealed.
With inner standard taper joint, bent 90° to the main chamber. Fabricated from borosilicate glass.
Customers Who Bought This Also Bought