Interference Check Standard AB(ICS-AB) Solution, Specpure®, Thermo Scientific
Fournisseur: Thermo Scientific Chemicals
Certificats
A propos de cet article
Molecular Formula: Matrix: 5% HNO3/tr. tartaric acid
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00147272
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.