Tweezers, Angled Fine Style 51, Excelta Corp®
Fournisseur: Excelta
Certificats
A propos de cet article
The offset oblique tips offer excellent visibility under a scope.
- Precision points for work under a scope
- Offset tips offer excellent visibility
- Offset tips excellent for accessibility in tight places
The '51 Style' series 45° offset oblique angle micro-fine tipped tweezer is ideal for working under a scope. It is also available with our Ergo-Tweeze® soft cushioned 'ET' grips (10¹⁰ ohms/sq surface resistivity), which also protect the user from heat. There is also a 30° offset style available. We offer 2 different qualities. The higher the quality, the more precise the tips. They are offered in stainless steel (rust-resistant) or stainless anti-magnetic steel (anti-magnetic).
Customers Who Bought This Also Bought