Craig Recrystallization Tube, Microscale, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Microscale Craig tube for recrystallizing small quantities of reaction product.
The assembly consists of a glass outer tube, with non-uniform grind to purposely allow leakage, and a PTFE inner plug. The plug eliminates breakage normally experienced with glass plugs. Stated volumes indicate the capacity below the grind. Fabricated from borosilicate glass.
Ordering information: Tube is available complete including glass outer member and PTFE inner plug, or as individual components that must be purchased separately. A glass inner plug is also available in lieu of PTFE.
Customers Who Bought This Also Bought