Replacement scalpel blades, Excelta Corp®
Supplier: Excelta
Certificates
About this item
These scalpel blades are all stainless steel (AISI 440) except the 181-22 which is carbon steel.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.