Tangent Galvanometer
# de catalogue 470137-638
Fournisseur: VWR International
Spécifications
- Description:Tangent Galvanometer
- Cat. no.:470137-638
- Supplier No.:PH0811
Spécifications
A propos de cet article
This versatile galvanometer demonstrates how a current loop generates a magnetic field.
Demonstrate how a current loop generates a magnetic field with this versatile galvanometer. Includes one 12.7cm dia. acrylic hoop wrapped with two coils of copper wire - one coil with five turns, and the other coil with ten turns. Three binding posts allow connections to five, ten, or 15 turns and a compass.
- Five, Ten, or Fifteen Turns
- Versatile
- Acrylic Hoop
- Three Binding Posts
- Two Coils Of Copper Wire
The galvanometer includes one 12.7 cm diameter acrylic hoop wrapped with two coils of copper wire. One coil has five turns, and the other coil has ten.
Three binding posts allow connections to five, ten, or 15 turns and a compass.
Customers Who Bought This Also Bought