Glass Stirring Shafts, 19 mm, Ace Glass
Fournisseur: Ace Glass
Certificats
A propos de cet article
Polished glass stirring shafts, 19mm O.D
Available in solid and hollow styles, and in various lengths. Join PTFE stir blades to button type shafts, or join PTFE multi-blade, PTFE single blade, PTFE turbine, and PTFE multi-paddle agitators to either knob or drilled style shafts. Recommended for use with ACE mechanical seal and PTFE debris-free bearings. Use ACE Flex-Grip chucks to join the shaft to a stirring motor.
Customers Who Bought This Also Bought