Thermowell with [ST] Joint, Ace Glass
Fournisseur: Ace Glass
Certificats
A propos de cet article
Thermowell features a 14/35 [ST] joint for use with spherical or cylindrical reaction flasks.
The length measurement (A as illustrated) listed below indicates the distance below the joint to the bottom of the tube. All Thermowells are fabricated from borosilicate glass.
Customers Who Bought This Also Bought