Analytical Funnels, Polypropylene, Ace Glass
Fournisseur: Ace Glass
Certificats
A propos de cet article
Made of polypropylene, these analytical funnels take standard size filter papers
The body of the funnel is at an exact 60° angle, and the internal ribs are 58° angle for rapid filtration. The outside ribbing prevents air lock. Funnels may be autoclaved.
Customers Who Bought This Also Bought