REDISHIP Protector® XStream® Laboratory Hoods Customisable with REDISHIP SpillStopper™ Work Surfaces, Labconco®
Fournisseur: Labconco
Certificats
A propos de cet article
Popular models of Protector XStream Laboratory Hoods and supporting matching work surfaces (sold separately) are available from VWR® stock for immediate shipment
REDISHIP SpillStopper™ Work Surfaces are dished to contain spills and feature a left rear corner cutout and oval cup Sink with cover.
REDISHIP Protector® XStream® Fiberglass Hoods feature left and right-sided GFCI duplexes and provides excellent containment and energy savings. Each hood is factory prepared to accommodate up to eight service fixtures (sold separately). The hood's by-pass block design restricts airflow, allowing the REDISHIP Protector® XStream® hood to operate as a reduced air volume (RAV) or variable air volume (VAV) system with no modifications.
Ordering information: Base cabinets are sold separately. Choose from service fixture kits and other accessories for customization (sold separately). Fiberglass and Coated Steel Blowers in sizes matched to most installation requirements are also available (sold separately).
Spécifications
- Catalog No:
- 89260-058
- 89260-064
- 89260-060
- 89260-062
- Environmentally Preferable:
- Sustainable Materials, Energy Efficient
- Sustainable Materials, Energy Efficient
- Sustainable Materials, Energy Efficient
- Sustainable Materials, Energy Efficient