Lavatory Sink, Knee Valve, Eagle MHC
# de catalogue 71001-940
Fournisseur: Eagle Group
Spécifications
- Depth:8 7/8"
- Description:Sink with Knee Valve Operation
- Cat. no.:71001-940
- Exterior Dimensions:15.75×19.75"
- Supplier No.:W1916
Spécifications
A propos de cet article
Type 304 stainless steel sink bowl with marine edge.
- Wall mounted
Delivery: Includes deck mounted gooseneck faucet, knee valves, soap dispenser and basket drain.
Customers Who Bought This Also Bought