AdjusTable® Workstations, Stainless Steel Solid Top with Overshelves, Stainless Steel Base, Eagle MHC™
Fournisseur: Eagle Group
Certificats
A propos de cet article
16 gauge Type 304 brushed stainless steel top.
- Base features 1 stainless steel wire shelf and 1 stainless steel plated C-frame
- Worsurface height: 34.5"
Ordering information: Include two 63" stainless steel posts and two 33" stainless steel posts and two 14" wide stainless steel wire overshelves.
Customers Who Bought This Also Bought