Wire Gowning Rack, Wall Mount, with Hooks, Eagle MHC
Supplier: Eagle Group
Certificates
About this item
Wall mounted gowning rack incorporates wire shelving components that offer a more economical rack option for less demanding environments.
- Convenient for storage of lab coats, uniforms and garments
- Open wire construction facilitates laminar flow
Hooks on all four shelf sides are spaced on 1¹/₂" centers and are staggered 1" vertically hook-to-hook for ease of access.
Frequently Bought Together